missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoProtect RNA stabilization reagent for cells and tissues.

Product Code. p-8929001
Click to view available options
Quantity:
50 mL
250 mL
500 mL
Unit Size:
250mL
500mL
50mL
This item is not returnable. View return policy

Product Code. 16432185

Brand: Macherey-Nagel™ 740400.50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

RNA stabilization reagent for cells, human and animal tissues, bacteria, yeast, insects, plant tissue, buffy coat and leukocytes.

  • Collect the sample first, get the RNA later
  • Protect your cell and tissue samples from RNA degradation
  • Compatible with most RNA isolation methods, includes NucleoSpin and NucleoMagRNA kits as well as NucleoZOL

Spécification

Content And Storage ≤ 24 hours at 37°C, ≤ 7 days at 18 to 25°C, ≤ 1 month at 4°C, long term at -20°C or lower
Sample Type Human/Animal Cells/Tissues (Max. 5 mm dia.)
For Use With (Equipment) Safe, transport of stabilized cell and tissue samples without dry ice for subsequent RNA isolation
For Use With (Application) Preservation of RNA integrity within cell and tissue samples (downstream applications: qRT-PCR, RNAseq,array technology, etc.)
Purity Typical RIN after RNA isolation: 10 for cultured mammalian cells (e.g., HeLa); > 9 for mammalian
Quantity 50 mL
Form RNA Stabilization Reagent for Cells and Tissues
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis