missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoProtect RNA stabilization reagent for cells and tissues.

Product Code. 16432185
Click to view available options
Quantity:
50 mL
250 mL
500 mL
Unit Size:
250mL
500mL
50mL
This item is not returnable. View return policy

Product Code. 16432185

Brand: Macherey-Nagel™ 740400.50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

RNA stabilization reagent for cells, human and animal tissues, bacteria, yeast, insects, plant tissue, buffy coat and leukocytes.

  • Collect the sample first, get the RNA later
  • Protect your cell and tissue samples from RNA degradation
  • Compatible with most RNA isolation methods, includes NucleoSpin and NucleoMagRNA kits as well as NucleoZOL

Specifications

Content And Storage ≤ 24 hours at 37°C, ≤ 7 days at 18 to 25°C, ≤ 1 month at 4°C, long term at -20°C or lower
Sample Type Human/Animal Cells/Tissues (Max. 5 mm dia.)
For Use With (Equipment) Safe, transport of stabilized cell and tissue samples without dry ice for subsequent RNA isolation
For Use With (Application) Preservation of RNA integrity within cell and tissue samples (downstream applications: qRT-PCR, RNAseq,array technology, etc.)
Purity Typical RIN after RNA isolation: 10 for cultured mammalian cells (e.g., HeLa); > 9 for mammalian
Quantity 50 mL
Form RNA Stabilization Reagent for Cells and Tissues
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.