missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nucleoporin 107 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-93328
This item is not returnable.
View return policy
Description
Nucleoporin 107 Polyclonal specifically detects Nucleoporin 107 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Nucleoporin 107 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| nuclear pore complex protein Nup107, nucleoporin 107kDa, NUP84Nucleoporin Nup107,107 kDa nucleoporin | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 mcg/mg lysate | |
| P57740 | |
| NUP107 | |
| The immunogen recognized by this antibody maps to a region between residue 875 and 925 of human nucleoporin 107kDa using the numbering given in entry NP_065134.1 (GeneID 57122). | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 57122 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction