missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nucleoporin 107 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | Nucleoporin 107 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30231775
|
Novus Biologicals
NBP3-35312-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30231860
|
Novus Biologicals
NBP3-35312-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nucleoporin 107 Polyclonal antibody specifically detects Nucleoporin 107 in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| Nucleoporin 107 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS (pH 7.3), 50% glycerol | |
| 57122 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| nuclear pore complex protein Nup107, nucleoporin 107kDa, NUP84Nucleoporin Nup107,107 kDa nucleoporin | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 766-925 of human Nucleoporin 107 (NP_065134.1).,, Sequence:, HMNSVPQKPALIPQPTFTEKVAHEHKEKKYEMDFGIWKGHLDALTADVKEKMYNVLLFVDGGWMVDVREDAKEDHERTHQMVLLRKLCLPMLCFLLHTILHSTGQYQECLQLADMVSSERHKLYLVFSKEELRKLLQKLRESSLMLLDQGLDPLGYEIQL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title