missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nucleoporin 107 Antibody [Janelia Fluor™ 635], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-18854JF635
This item is not returnable.
View return policy
Description
Nucleoporin 107 Polyclonal antibody specifically detects Nucleoporin 107 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Nucleoporin 107 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Janelia Fluor 635 | |
| nuclear pore complex protein Nup107, nucleoporin 107kDa, NUP84Nucleoporin Nup107,107 kDa nucleoporin | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Nucleoporin 107 | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 57122 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido