missing translation for 'onlineSavingsMsg'
Learn More

Nucleoporin 107 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication

£141.00

Specifications

Antigen Nucleoporin 107
Concentration 1 mg/mL
Dilution Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10 μg/mL
Applications Western Blot
Classification Polyclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18574954
View Documents
Novus Biologicals
NBP1-76926-0.025mg
0.025 mg
£141.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Nucleoporin 107 Polyclonal specifically detects Nucleoporin 107 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Nucleoporin 107
Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10 μg/mL
Polyclonal
Rabbit
Core ESC Like Genes, Stem Cell Markers
PBS with 0.02% Sodium Azide
nuclear pore complex protein Nup107, nucleoporin 107kDa, NUP84Nucleoporin Nup107,107 kDa nucleoporin
NUP107
IgG
Affinity Purified
1 mg/mL
Western Blot
Unconjugated
RUO
Human
P57740
57122
Antibody was raised against a 20 amino acid synthetic peptide from near the center of human NUP107. The immunogen is located within amino acids 370 - 420 of NUP107. Amino Acid Squence: HDPNVNGGTELEPVEGNPYR
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.