missing translation for 'onlineSavingsMsg'
Learn More

Nucleoporin 107 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18408172 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408172 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408172 Supplier Novus Biologicals Supplier No. NBP176926AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Nucleoporin 107 Polyclonal antibody specifically detects Nucleoporin 107 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nucleoporin 107
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias nuclear pore complex protein Nup107, nucleoporin 107kDa, NUP84Nucleoporin Nup107,107 kDa nucleoporin
Host Species Rabbit
Immunogen Antibody was raised against a 20 amino acid synthetic peptide from near the center of human NUP107. The immunogen is located within amino acids 370 - 420 of NUP107. Amino Acid Squence: HDPNVNGGTELEPVEGNPYR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 57122
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.