missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Nucleoplasmin-3 Antibody, Novus Biologicals™

Product Code. 18347669 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18347669 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18347669 Supplier Novus Biologicals™ Supplier No. H00010360B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

Nucleoplasmin-3 Polyclonal antibody specifically detects Nucleoplasmin-3 in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nucleoplasmin-3
Applications Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunocytochemistry/ Immunofluorescence 1:10-1:500
Formulation PBS (pH 7.4)
Gene Accession No. NP_008924.1
Gene Alias nucleophosmin/nucleoplasmin 3, nucleophosmin/nucleoplasmin family, member 3, nucleoplasmin-3, PORMIN, TMEM123
Host Species Mouse
Immunogen NPM3 (NP_008924.1, 1 a.a. - 178 a.a.) full-length human protein. MAAGTAAALAFLSQESRTRAGGVGGLRVPAPVTMDSFFFGCELSGHTRSFTFKVEEEDDAEHVLALTMLCLTEGAKDECNVVEVVARNHDHQEIAVPVANLKLSCQPMLSLDDFQLQPPVTFRLKSGSGPVRITGRHQIVTMSNDVSEEESEEEEEDSDEEEVELCPILPAKKQGGRP
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10360
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.