missing translation for 'onlineSavingsMsg'
Learn More

Nucleoplasmin-2 Antibody (5E9), Novus Biologicals™

Product Code. 18334029 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18334029 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18334029 Supplier Novus Biologicals Supplier No. H00010361M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Nucleoplasmin-2 Monoclonal antibody specifically detects Nucleoplasmin-2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nucleoplasmin-2
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 5E9
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_877724.1
Gene Alias MGC78655, nucleophosmin/nucleoplasmin 2, nucleoplasmin-2
Host Species Mouse
Immunogen NPM2 (NP_877724.1, 1 a.a. ~ 214 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MNLSSASSTEEKAVTTVLWGCELSQERRTWTFRPQLEGKQSCRLLLHTICLGEKAKEEMHRVEILPPANQEDKKMQPVTIASLQASVLPMVSMVGVQLSPPVTFQLRAGSGPVFLSGQERYEASDLTWEEEEEEEGEEEEEEEEDDEDEDADISLEEQSPVKQVKRLVPQKQASVAKKKKLEKEEEEIRASVRDKSPVKKAKATARAKKPGFKK
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Developmental Biology
Primary or Secondary Primary
Gene ID (Entrez) 10361
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.