missing translation for 'onlineSavingsMsg'
Learn More

Nucleophosmin (Acute Myeloid Leukemia Marker) Mouse, Clone: NA24, enQuireBio™

Mouse Monoclonal Antibody

£181.83 - £423.29

Specifications

Antigen Nucleophosmin (Acute Myeloid Leukemia Marker)
Clone NA24
Concentration 200μg/mL
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15930598
View Documents
EnquireBio
4869-MSM3-P0
20 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15940598
View Documents
EnquireBio
4869-MSM3-P1
100 μg
£423.29
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Nucleophosmin (Acute Myeloid Leukemia Marker)
200μg/mL
Monoclonal
Mouse
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide.
GST fusion protein containing the N-terminal part of nucleophosmin fused to 14 amino acid of ALK protein
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
33kDa
NA24
Immunohistochemistry
Unconjugated
RUO
4869
IgG1 κ
Protein A/G purified from Bioreactor concentrate.
Nuclear and Cytoplasmic
MCF-7, HeLa or A431 cells. Skin, Colon
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.