missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Includes
Reagents A and B, 5.0M Sodium Perchlorate, and Nucleon resins.
- Phenol-free protocol requiring only 30 minutes to complete
- Low-shearing protocol gives maximum recoveries of high-molecular weight DNA
- For 50 preps of 1mL of whole blood or cultured cells (1 to 3 x 106 cells/mL)
- Available in different formats/reaction scales (See 45-000-956 and 45-000-960)
Specifications
Specifications
| Description | illustra Nucleon BACC system rapidly extracts high molecular weight DNA from whole blood and cultured cells. |
| Sample Type | Blood, Cell Cultures |
| For Use With (Application) | Rapid extraction of high-quality, high molecular weight genomic DNA from cell cultures and blood. |
| Includes | Reagents A and B, 5M Sodium Perchlorate and Nucleon™ Resins |
| Product Type | illustra Nucleon BACC1 |
| Quantity | 25 Kits |
| Yield | < 12 μg DNA/mg tissue |
Safety and Handling
Signal Word
- Danger
- Oxidising liquid Category 2
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501-Dispose of contents/container to .
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?