missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoMag Tissue Kit (DNA Purification from Cells and Tissue)

Product Code. 18854360
Change view
Click to view available options
Quantity:
384 Preps
96 Preps
Unit Size:
Pack of 96
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18854360 384 Preps -
18844360 96 Preps Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18854360 Supplier Macherey-Nagel™ Supplier No. 744330.4
  • ONLINE EXCLUSIVE PRICE
    £1420.01
    £1292.24 / Pack of 384
    Save £127.77
    8% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Streamlined magnetic-bead DNA purification from a wide range of cell and tissue samples using the NucleoMag Tissue Kit.

  • The NucleoMag Tissue kit combines the flexibility and versatility of the Tissue product series with magnetic bead-based processing.
  • Provides an automation-friendly workflow for DNA purification.
  • Uses a simple protocol that is effective for a broad range of sample types.
  • Compatible with a variety of open robotic platforms.

Specifications

Capacity 0.4 μg/μL Beads
Isolation Technology Magnetic Bead
Sample Type Cells, Human Tissue, Animal Tissue
Elution Volume 50 to 200 μL
Final Product Type DNA
For Use With (Application) Isolation of DNA
Product Line NucleoMag
Product Type NucleoMag Tissue Kit
Quantity 384 Preps
Purity 1.6 to 1.9
Sample Volume <20 mg Human/Animal Tissue, <106 Cells
Technique Magnetic Separation
Test Time 40 to 120 min./96 Preps
Yield 10 to 20 μg (20 mg tissue)
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • NucleoMag DNA Microbiome (4x96)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: triethanolamine, 2-propanol, guanidine hydrochloride, ethanol, EDTA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.