missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoMag SEP Magnetic Separators
Click to view available options
Product Type:
SEP 24
SEP Maxi
SEP Mini
For Use With (Equipment):
1.5 / 2.0 mL tubes
24-well plate
50 mL tubes
No. of Samples:
4
12
24
Unit Size:
Each
Description
- NucleoMag™ kits are based on magnetic-bead technology and allow rapid simultaneous purification of high-quality nucleic acids
- NucleoMag™ Beads are attracted to the magnet in the separator, which allows removal of the wash buffer or recovery of the elution buffer containing purified DNA
- Consists of a polytetrafluorethylen (PTFE) base and magnetic pins
- Resistant to diluted acetic acid, ethanol, guanidine-HCl, NaCl, NaOH, and SDS
- Avoid prolonged exposure to chemicals; do not autoclave
Specifications
Specifications
| Content And Storage | 1 Ea., 15°C to 25°C |
| Description | PTFE base, Magnetic pins |
| Detection Method | Magnetic |
| For Use With (Equipment) | 1.5 / 2.0 mL tubes |
| For Use With (Application) | Nucleic acid separation using NucleoMag™ kits |
| No. of Samples | 12 |
| Product Type | SEP Mini |
| Quantity | 1 Ea. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction