missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoMag RNA kit for magnetic bead based RNA purification

Product Code. 12883931
Change view
Click to view available options
No. of Reactions:
4 x 96
96
Unit Size:
384 reactions
96 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
12883931 96 96 reactions
12757790 4 x 96 384 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12883931 Supplier Macherey-Nagel™ Supplier No. 744350.1
  • ONLINE EXCLUSIVE PRICE
    £387.00
    £331.91 / 96 reactions
    Save £55.09
    14% Off
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Isolate RNA through a magnetic-bead based platform. Macherey Nagel™ NucleoMag™ RNA Kit enables isolation of RNA from tissue and cell samples.

Features
  • One-tube procedure minimizes risk of cross-contamination
  • Small elution volumes ≥ 50μL
  • Suitable for manual and automated processing
  • Recombinant Dnase included
  • Reducing agent TCEP included (no β-mercaptoethanol necessary)
Applications
  • Rapid manual and automated small-scale preparation of highly pure total RNA from tissue or cell samples

Specifications

Format Highly Reactive Superparamagnetic Beads
For Use With (Application) Rapid manual and automated small-scale preparation of highly pure total RNA from tissue or cell samples
Includes NucleoMag™ B-Beads, buffers, TCEP, RNase-free rDNase
No. of Reactions 96
Product Type NucleoMag RNA
RNase Yes
Sample Size <2 x 106 cells, <20 mg human/animal tissue
Test Time <120 min./96 Reaction
Yield <30μg
compliance-icons compliance-icons compliance-icons
Product Identifier
  • NucleoMag RNA
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • LONG-TERM AQUATIC HAZARD Chronic 4
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H412-Harmful to aquatic life with long lasting effects.
  • H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: guanidinium thiocyanaten,ethanol/tris-(2-carboxyethyl)phosphine hydrochloride, TCEP(
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.