missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoMag DNA Swab for DNA purification from swabs

Product Code. 15880978
Click to view available options
No. of Reactions:
1 x 96
24 x 96
4 x 96
Unit Size:
192 reactions
2304 reactions
96 reactions
This item is not returnable. View return policy
UN 1993 Flammable liquid, n.o.s. (ethanol mixture) Chem class 3 Product group II

Product Code. 15880978

Brand: Macherey-Nagel™ 744601.24

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
UN 1993 Flammable liquid, n.o.s. (ethanol mixture) Chem class 3 Product group II

Magnetic bead based kit for the isolation of genomic DNA from swabs.

  • High throughput DNA isolation for genetic testing
  • Developed for cotton as well as synthetic swabs
  • Combine with NucleoSpin™ Forensic Filters for convenient sample prep

Specifications

Capacity 0.4 μg/μL beads
Concentration 10 to 30 ng/μL
Isolation Technology Magnetic Bead
Sample Type Swab (Cotton or Synthetic)
Efficiency 0.4 μg/μL beads theoretical binding capacity
For Use With (Equipment) COPAN FLOQSwabs™, Puritan HydraFlock™ swabs
For Use With (Application) Typical downstream applications: qPCR, NGS, STR analysis, any kind of enzymatic reaction
No. of Reactions 24 x 96
Purity A260/A280: 1.6 - 1.9
Test Time 30 min/ 96 preps on KingFisher™ Flex (excluding lysis)
Form Highly Reactive Superparamagnetic Beads
Show More Show Less

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.