missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoMag cfDNA kit for cell-free DNA purification from plasma
Click to view available options
No. of Reactions:
4 x 48 Preps
48 Preps
Unit Size:
192 reactions
48 reactions
Description
NucleoMag™ DNA Plasma
- Consistent circulating cell-free recovery from 1–10 mL plasma samples
- Manual and automated isolation
- Efficient purification of fragmented DNA as small as 50 bp
- No PCR inhibition regardless of your preferred sample volume
- Human plasma samples derived from EDTA blood draw tubes or Cell-Free DNA BCT
- For Liquid Biopsy analysis, NGS, qPCR, ddPCR
Specifications
Specifications
| Capacity | 0.3 μg/μL beads |
| Content And Storage | Room temperature (18 - 25 °C) |
| Isolation Technology | Magnetic Bead |
| Sample Type | Human EDTA/Cell-Free DNA BCT™ Plasma |
| Efficiency | Binding capacity: 0.3 μg/μL beads |
| Elution Volume | Elution volume: 50 - 200 μL |
| For Use With (Equipment) | Magnetic separator, separation plate |
| For Use With (Application) | Isolation of cell-free DNA from plasma |
| Includes | NucleoMag P-Beads, buffers, Liquid Proteinase K |
| No. of Reactions | 4 x 48 Preps |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction