missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoMag cfDNA kit for cell-free DNA purification from plasma
Klicka för att se tillgängliga alternativ
No. of Reactions:
4 x 48 Preps
48 Preps
Förpackningsstorlek:
192 reactions
48 reactions
Beskrivning
NucleoMag™ DNA Plasma
- Consistent circulating cell-free recovery from 1–10 mL plasma samples
- Manual and automated isolation
- Efficient purification of fragmented DNA as small as 50 bp
- No PCR inhibition regardless of your preferred sample volume
- Human plasma samples derived from EDTA blood draw tubes or Cell-Free DNA BCT
- For Liquid Biopsy analysis, NGS, qPCR, ddPCR
Specifikationer
Specifikationer
| Capacity | 0.3 μg/μL beads |
| Content And Storage | Room temperature (18 - 25 °C) |
| Isolation Technology | Magnetic Bead |
| Sample Type | Human EDTA/Cell-Free DNA BCT™ Plasma |
| Efficiency | Binding capacity: 0.3 μg/μL beads |
| Elution Volume | Elution volume: 50 - 200 μL |
| For Use With (Equipment) | Magnetic separator, separation plate |
| For Use With (Application) | Isolation of cell-free DNA from plasma |
| Includes | NucleoMag P-Beads, buffers, Liquid Proteinase K |
| No. of Reactions | 48 Preps |
| Visa mer |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering