missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoMag DNA Food kit for DNA purification from food

Product Code. p-7159637
Click to view available options
Quantity:
384 preps
96 preps
Unit Size:
Each
Pack of 4
This item is not returnable. View return policy

Product Code. 15865224

Brand: Macherey-Nagel™ 744945.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Flexible magnetic bead based isolation of DNA from food and feed.

NucleoMag™ DNA Food kits

  • DNA extraction from food and feed for e.g., species identification, GMO detection
  • Get even low amounts of partially degraded DNA from complex matrices
  • Extraction of DNA from contaminating bacteria (food safety)
  • Kit chemistry allows full sample flexibility
  • Manual or automated isolation of DNA
  • Typical downstream application: real-time PCR

Specifications

Capacity 0.4 μg/μL beads
Content And Storage Room temperature (18 - 25 °C)
Isolation Technology Magnetic Bead
Sample Type Feed, Food
Efficiency Binding capacity: 0.4 μg/μL beads
Elution Volume Elution volume: 50 - 200 μL
For Use With (Equipment) Magnetic separator
For Use With (Application) Isolation of DNA from food and feed
Includes NucleoMag B-Beads, buffers, Proteinase K
No. of Samples 1 x 96
Quantity 96 preps
Purity A260/A280: 1.6 - 1.9
Sample Volume <200 mg
Test Time 40 - 120 min/96 preps (excl. lysis, depending on instrument)
Yield 0.1 - 10 μg
Form Highly Reactive Superparamagnetic Beads
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • NucleoMag DNA Food (1x96)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Supplemental information
  • MIXTURE LIST-Contains : guanidium hydrochloride, 2-propanol, EDTA, ethanol, tris(hydroxymethyl) aminomethane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.