missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoMag DNA Food kit for DNA purification from food

Product Code. 15865224
Change view
Click to view available options
Quantity:
384 preps
96 preps
Unit Size:
Each
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15865224 96 preps Each
15875224 384 preps Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15865224 Supplier Macherey-Nagel™ Supplier No. 744945.1
  • ONLINE EXCLUSIVE PRICE
    £301.00
    £248.25 / Pack of 1
    Save £52.75
    17% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Flexible magnetic bead based isolation of DNA from food and feed.

NucleoMag™ DNA Food kits

  • DNA extraction from food and feed for e.g., species identification, GMO detection
  • Get even low amounts of partially degraded DNA from complex matrices
  • Extraction of DNA from contaminating bacteria (food safety)
  • Kit chemistry allows full sample flexibility
  • Manual or automated isolation of DNA
  • Typical downstream application: real-time PCR

Spezifikation

Capacity 0.4 μg/μL beads
Content And Storage Room temperature (18 - 25 °C)
Isolation Technology Magnetic Bead
Sample Type Feed, Food
Efficiency Binding capacity: 0.4 μg/μL beads
Elution Volume Elution volume: 50 - 200 μL
For Use With (Equipment) Magnetic separator
For Use With (Application) Isolation of DNA from food and feed
Includes NucleoMag B-Beads, buffers, Proteinase K
No. of Samples 1 x 96
Quantity 96 preps
Purity A260/A280: 1.6 - 1.9
Sample Volume <200 mg
Test Time 40 - 120 min/96 preps (excl. lysis, depending on instrument)
Yield 0.1 - 10 μg
Form Highly Reactive Superparamagnetic Beads
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons
Produktidentifikator
  • NucleoMag DNA Food (1x96)
Signalwort
  • Warning
Gefahrenkategorie
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Ergänzende Informationen
  • MIXTURE LIST-Contain: guanidium hydrochloride, 2-propanol, EDTA, ethanol, tris(hydroxymethyl) aminomethane
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.