missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoMag Clearing Beads - Magnetic Beads for Lysate Clarification

Product Code. 30694921
Change view
Click to view available options
Isolation Technology:
Beads
Unit Size:
96 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Isolation Technology unitSize
30694921 Beads 96 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 30694921 Supplier Macherey-Nagel™ Supplier No. 744751.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

The NucleoMag Clearing Beads can be used for lysate clarification during plasmid DNA isolation with the NucleoMag Plasmid kit.

  • They offer a simple solution for lysate clarification on magnetic rod systems and can replace centrifugation.

Specifications

Cell Type Bacteria,E. coli
Content And Storage 3.5 mL
Format Beads
Isolation Technology Beads
Sample Type Bacteria
Final Product Type Plasmid DNA
For Use With (Application) High copy plasmid DNA purification
No. of Reactions 3.5 mL
Product Line NucleoMag
Product Type Magnetic Beads
Quantity 3.5 mL
Sample Volume 0.5 to 1.5 mL E.coli Culture
Technique Magnetic separation
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.