missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoMag VET kit for viral/bacterial DNA and viral RNA from veterinary samples
Click to view available options
No. of Reactions:
1 x 96 Preps
4 x 96
Unit Size:
1 set
96 reactions
Specifications
Specifications
| Capacity | 0.4 μg/μL beads |
| Final Product Type | Nucleo Mag VET |
| No. of Reactions | 1 x 96 Preps |
| Probe Length | 300 to 50bp to kbp |
| Product Type | Nucleo Mag VET |
| Sample Volume | <200 μL whole blood/serum/plasma/swab solution/feces, 10 to 30 mg tissue |
Safety and Handling
Signal Word
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- LONG-TERM AQUATIC HAZARD Chronic 3
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contains : ethanol,sodium perchlorate / guanidine hydrochloride
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction