missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoMag VET kit for viral/bacterial DNA and viral RNA from veterinary samples

Product Code. 15301204
Click to view available options
No. of Reactions:
1 x 96 Preps
4 x 96
Unit Size:
1 set
96 reactions
This item is not returnable. View return policy

Product Code. 15301204

Brand: Macherey-Nagel™ 744200.4

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Magnetic bead based kit for isolation of viral nucleic acids and bacterial DNA from veterinary samples.

Specifications

Capacity 0.4 μg/μL beads
Final Product Type Nucleo Mag VET
No. of Reactions 4 x 96
Probe Length 300 to 50bp to kbp
Product Type Nucleo Mag VET
Sample Volume <200 μL whole blood/serum/plasma/swab solution/feces, 10 to 30 mg tissue
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : ethanol,sodium perchlorate / guanidine hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.