missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoMag Virus kit for RNA/DNA purification from cell-free fluids

Product Code. 12738682
This item is not returnable. View return policy

Product Code. 12738682

Brand: Macherey-Nagel™ 744800.4

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Magnetic bead based kit for isolation of viral RNA and DNA from biological fluids.

4 x 96 well plate

Specifications

Capacity 0.2 μg/μL beads
Content And Storage 18°C to 25°C
Sample Type Cell-free Biological Fluids, Plasma, Serum
For Use With (Application) Isolation of Viral Nucleic Acids
No. of Reactions 384 Preps
Sample Volume <200 μL
Size 300 bp−approx. 50 kbp
Target Viral RNA and DNA
Form Magnetic Beads
compliance-icons compliance-icons compliance-icons
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.