missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel Bioanalysis™ NucleoMag 96 PCR Kit with Magnetic Beads

Product Code. 12783580
Change view
Click to view available options
Quantity:
384
Unit Size:
384 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12783580 384 384 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12783580 Supplier Macherey-Nagel Bioanalysis™ Supplier No. 744100.4

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Magnetic bead based PCR clean up for highest flexibility.

The NucleoMag 96 PCR procedure is based on reversible adsorption of nucleic acids to paramagnetic beads under appropriate buffer conditions. Adjusting the PCR reaction to binding conditions and addition of paramagnetic beads can be carried out simultaneously. After magnetic separation and removal of supernatant, the beads are washed to remove contaminants and salt. A short drying step is necessary to remove ethanol from previous washing steps. Finally, highly purified DNA is eluted with low salt elution buffer or water and can be used directly for downstream applications. The NucleoMag 96 PCR kit can be used either manually or automated on standard liquid handling instruments.

  • Magnetic bead technology
  • Sample Amount: < 50 μL
  • Fragment Size: 150 bp to 10 kbp
  • Typical Recovery: 80 to 95 %
  • Binding Capacity: 0.3 μg/μL beads
  • Preparation Time: 40 to 120 min/96 preps
  • Storage: 18 to 25°C

Specifications

Sample Type PCR Reaction Mixture
Elution Volume (Metric) 25 to 100 μL
For Use With (Application) PCR clean up
Quantity 384
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Supplemental information
  • MIXTURE LIST-Contain: guanidine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.