missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoMag 384 Plant kit for DNA purification from plants

Product Code. 16371626
Change view
Click to view available options
No. of Reactions:
1 x 384
4 x 384
Unit Size:
1536 reactions
384 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
16371626 4 x 384 1536 reactions
16361626 1 x 384 384 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
UN 1993 Flammable liquid, n.o.s. (2-propanol, ethanol mixture) Chem class 3 Product group II
Product Code. 16371626 Supplier Macherey-Nagel™ Supplier No. 744402.4
  • ONLINE EXCLUSIVE PRICE
    £935.00
    £771.11 / 1536 reactions
    Save £163.89
    17% Off
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
UN 1993 Flammable liquid, n.o.s. (2-propanol, ethanol mixture) Chem class 3 Product group II

Flexible magnetic bead based isolation of DNA from a multitude of plant samples.

  • Optimized for high throughput processing in a 384-well format
  • Efficient purification of DNA from up to 30 mg sample material
  • Suitable for a wide variety of plant species
  • Typical downstream applications: PCR, Southern blotting, any kind of enzymatic reaction

Specifications

Capacity 0.2 μg/μL beads
Isolation Technology Magnetic Bead
Sample Type Plant Tissue
Efficiency 0.2 μg/μL beads Binding capacity
No. of Reactions 4 x 384
Sample Volume <30 mg wet weight
Test Time 40-120 min/ 96 preps (excl. sample lysis); 60 min/ 384 preps (excl. sample lysis)
Yield Depending on sample source, storage and quality
Form Highly Reactive Superparamagnetic Beads

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.