missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Nucleolin Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA519508
This item is not returnable.
View return policy
Description
Heat mediated antigen retrieval recommended prior to tissue staining. Store antibody at 4ºC for 1-2 weeks. For long-term storage, store at -20ºC.
Nucleolin (NCL), a eukaryotic nucleolar phosphoprotein, is involved in the synthesis and maturation of ribosomes. It is located mainly in dense fibrillar regions of the nucleolus. Human NCL gene consists of 14 exons with 13 introns and spans approximately 11kb. The intron 11 of the NCL gene encodes a small nucleolar RNA, termed U20.
Specifications
| Nucleolin | |
| Polyclonal | |
| Unconjugated | |
| NCL | |
| B530004O11Rik; C23; D0Nds28; D1Nds28; FLJ45706; FLJ59041; Ncl; Nuc; Nucl; nucl antibody; Nucleolin; nucleolin-like protein; Protein C23 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 17975, 25135, 4691 | |
| -20°C or -80°C if preferred | |
| Liquid |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide; pH 7.4 | |
| P09405, P13383, P19338 | |
| NCL | |
| Synthetic peptide conjugated to KLH derived from within residues 1 - 100 of Human Nucleolin. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction