missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nucleolin Antibody (NCL/8068R), Novus Biologicals™
Rabbit Monoclonal Antibody
£205.00 - £377.00
Specifications
| Antigen | Nucleolin |
|---|---|
| Clone | NCL/8068R |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot : 1-2 μg/mL, Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18633034
|
Novus Biologicals
NBP3-23668-20ug |
20 μg |
£205.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18611365
|
Novus Biologicals
NBP3-23668-100ug |
100 μg |
£377.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nucleolin Monoclonal antibody specifically detects Nucleolin in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
| Nucleolin | |
| 0.2 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Phospho Specific, Stem Cell Markers | |
| 10 mM PBS with 0.05% BSA | |
| 4691 | |
| IgG κ | |
| Protein A or G purified |
| NCL/8068R | |
| Western Blot : 1-2 μg/mL, Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| C23, FLJ45706, FLJ59041, nucleolin, Protein C23 | |
| Recombinant full-length human Nucleolin protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title