missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoCard for blood storage at room temperature

Product Code. 11962512
Click to view available options
Quantity:
10 Pieces
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 11962512

Brand: Macherey-Nagel™ 740403.10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Blood sample storage card for subsequent DNA extraction.

Specifications

Content And Storage 18°C to 25°C
Format Filter Card
Sample Type Blood
For Use With (Application) Blood storage for DNA extraction
Quantity 10 Pieces
Sample Volume <200 μL
Target Auxiliary tools
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.