missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nucleocapsid protein Human HEK293 anti-SARS-CoV-2, Unconjugated, Clone: AS41, AcroBiosystems
Human HEK293 Monoclonal Antibody
£374.00 - £2730.00
Specifications
| Antigen | Nucleocapsid protein |
|---|---|
| Clone | AS41 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Host Species | Human (HEK293) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17106571
|
ACROBiosystems
NUN-S41-100ug |
100 μg |
£374.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17116571
|
ACROBiosystems
NUN-S41-1mg |
1 mg |
£2730.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nucleocapsid protein Monoclonal antibody specifically detects Nucleocapsid protein in SARS-CoV-2 samples.Specifications
| Nucleocapsid protein | |
| Monoclonal | |
| Human (HEK293) | |
| SARS-CoV-2 | |
| Primary |
| AS41 | |
| Unconjugated | |
| RUO | |
| IgG1 | |
| -20°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title