missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoBond™ Xtra Maxi Plus kit for transfection-grade plasmid DNA

Product Code. 12758412
Change view
Click to view available options
No. of Reactions:
2
10
Unit Size:
10 reactions
2 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
12758412 10 10 reactions
11915452 2 2 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12758412 Supplier Macherey-Nagel™ Supplier No. 740416.10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Purify transfection-grade plasmid DNA with high yield. The NucleoBond™ Xtra Maxi Plus Kit represents the new generation of anion exchanger allowing high yield, ultra-fast. The Plus kit contains NucleoBond™ Finalizers to speed up DNA precipitation.

Features
  • Proven anion-exchange technology – transfection-grade plasmid DNA
  • Patented endotoxin removal by additional washing step – no extra ice incubation necessary
  • Optimized column design – Maxi prep in 35 min.
  • Column filter included – high filter flow-rates, parallel lysate clearing and loading onto the column
  • Improved silica material – typical yields of 1000μg (Maxi)
  • NucleoBond™ Xtra EF kits contain NucleoBond™ Finalizers to speed up DNA precipitation
Applications
  • High-copy plasmids from E. coli
  • Low-copy plasmids from E. coli (increased buffer volumes required)

Specifications

Buffer Yes
Format Maxi Gravity-flow Column
Isolation Technology Anion Exchange Chromatography Technology
No. of Reactions 10
RNase Yes
Sample Volume <600 mL (high copy), <1200 mL (low copy)
Size <300 kbp
Yield 1000μg
compliance-icons compliance-icons compliance-icons
Product Identifier
  • NucleoBond Xtra Maxi Plus
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: 2-propanol/sodium hydroxide solution (diluted < 2 %),dodecylsulfate, sodium salt/ethanol (diluted <
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.