missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoBond™ Xtra Maxi kit for transfection-grade plasmid DNA

Product Code. 11902312
Click to view available options
No. of Reactions:
10
50
100
Unit Size:
0.1kg
100 preps
50 preps
This item is not returnable. View return policy

Product Code. 11902312

Brand: Macherey-Nagel™ 740414.100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Purify transfection-grade plasmid DNA with high yield. The NucleoBond™ Xtra Maxi Kit represents the new generation of anion exchanger. The optimized design results in high yield and ultra-fast processing.

Features

  • Optimized column design—Silica resin packed between filters, with prep in just 75 minutes
  • Column filter included—High filter flow rates, parallel lysate clearing, and loading onto the column
  • Improved silica material—Typical yields of 1,000μg and can be used over a wide pH range
  • Proven anion-exchange technology—Transfection-grade plasmid DNA
  • Kits contain: NucleoBond™ Xtra Maxi columns and filter, RNase A, buffers (RES, LYS, NEU, EQU, WASH, ELU) and plastic washers

Applications

  • High-copy plasmids from E. coli
  • Low-copy plasmids from E. coli (increased buffer volumes required)

Specifications

Buffer Yes
Format Maxi Gravity-flow Column
Isolation Technology Anion Exchange Chromatography Technology
No. of Reactions 100
Purity A260/A280:1.80–1.95
RNase Yes
Sample Volume <600 mL (high copy), <1200 mL (low copy)
Size <300 kbp
Test Time 75 min./prep
Yield 1000μg
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : 2-propanol/sodium hydroxide solution (diluted < 2 %),dodecylsulfate, sodium salt/ethanol (diluted <
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.