missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoBond™ Xtra Maxi Columns
Click to view available options
:
No. of Reactions: 100
Description
NucleoBond™ Xtra—The new generation of anion exchanger for ultra-fast transfection-grade plasmid DNA
- Optimized column design – Maxi prep in 35 min
- Typical yield of 1000 μg (Maxi)
- Polypropylene column suitable for use with organic solvents and buffers with denaturing agents
- Column filter included for high filter flow rates, parallel lysate clearing, and loading onto the column
- Depth filters avoid time-consuming centrifugation for lysate clearing
- Complete removal of precipitate even with large lysate volumes without clogging
- Avoids shearing of large DNA constructs (such as PACs or BACs)
Specifications
Specifications
| Format | Separate Columns |
| Isolation Technology | Anion Exchange Chromatography Technology |
| Sample Type | E. coli Culture |
| Final Product Type | Transfection Grade Plasmid DNA |
| For Use With (Equipment) | NucleoBond™ Xtra Maxi Kits |
| For Use With (Application) | Plasmid DNA Purification |
| No. of Reactions | 100 Reactions |
| Product Type | Gravity Flow column |
| Purity | Transfection Grade |
| Sample Volume | < 200 mL (high copy), <400 mL (low copy) |
| Show More |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction