missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoBond™ Xtra Elution Buffer

Product Code. 16361606
Change view
Click to view available options
Quantity:
900 mL
Unit Size:
900mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16361606 900 mL 900mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
UN 1993 Flammable liquid, n.o.s. (2-propanol mixture) Chem class: 3 Pack group: III
Product Code. 16361606 Supplier Macherey-Nagel™ Supplier No. 740316.900

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
UN 1993 Flammable liquid, n.o.s. (2-propanol mixture) Chem class: 3 Pack group: III

ELU Buffer for Plasmid DNA purification

Specifications

For Use With (Application) Plasmid DNA Purifcation
For Use With (Equipment) NucleoBond™ Xtra Plasmid Kits
Physical Form Buffer
Quantity 900 mL
compliance-icons compliance-icons
Product Identifier
  • Buffer ELU (900 mL)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contain: propan-2-ol

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.