missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoBond™ Xtra EF Buffer Set I
Description
NucleoBond™ Xtra EF s a new generation of anion exchangers for isolation of endotoxin-free, low-copy plasmids, cosmids, BACs, PACs and P1 constructs. It features ultra-fast filtration and high yield, with a patented endotoxin removal process that required no extra ice incubation.
- NucleoBond™ Xtra EF kits include resuspension, cell lysis and neutralization buffers, plus DNase-free RNase
- When a larger amount of low-copy plasmid is required, larger culture volumes can be prepared using additional buffer volumes
- Buffer Set I provides 150 mL of buffers RES-EF, LYS-EF, NEU-EF and RNase A for use only with NucleoBond™ Xtra EF Kits
Specifications
Specifications
| Chemical Name or Material | NucleoBond™ Xtra EF Buffer Set I |
| For Use With (Application) | Isolation of Endotoxin-Free Low-Copy Plasmids, Cosmids, BACs,PACs, and P1 Constructs |
| For Use With (Equipment) | NucleoBond™ Xtra EF Midi, Maxi, Midi Plus and Maxi Plus Kits |
| Quantity | Sufficient for 10 Xtra EF Maxi or 20 Xtra EF Midi preps |
| Includes | 150mL each of:
|
Safety and Handling
Signal Word
- Danger
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?