missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoBond™ Xtra Buffer Set I

Product Code. 12810931
Change view
Click to view available options
Quantity:
10 Xtra Maxi or 20 Xtra Midi Preps
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12810931 10 Xtra Maxi or 20 Xtra Midi Preps 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12810931 Supplier Macherey-Nagel™ Supplier No. 740417

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Provides additional buffer for use when larger culture volumes are needed. Macherey-Nagel™ NucleoBond™ Xtra Buffer Set I includes 150mL of buffers RES, LYS, NEU, and RNase A, sufficient for 10 Xtra Maxi preps or 20 Xtra Midi preps.

NucleoBond™ Xtra is a new generation of anion exchangers featuring fast filtration, high binding capacity and high flow rate. They yield ultra-pure plasmid DNA in about half the time compared with other Midi or Maxi kits based on anion-exchange chromatography.

  • NucleoBond™ Xtra kits include resuspension, cell lysis and neutralization buffers, plus DNase-free RNase
  • Some applications (e.g. low-copy vector purification) require a larger culture volume, which necessitates a corresponding increase in buffer volumes 
  • Buffer Set I provides 150mL of buffers RES, LYS, NEU and RNase A for use only with NucleoBond™ Xtra Kits

Specifications

Chemical Name or Material NucleoBond™ Xtra Buffer Set I
For Use With (Application) Plasmid DNA purification
For Use With (Equipment) NucleoBond™ Xtra Midi, Maxi, Midi Plus and Maxi Plus Kits
Quantity 10 Xtra Maxi or 20 Xtra Midi Preps
Includes 150 mL each of: Buffer RES, Buffer LYS, Buffer NEU<, RNase A
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: sodium hydroxide, dodecyl sulphate, RNAse
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.