Learn More
Macherey-Nagel™ NucleoBond™ Smart Rack
Description
NucleoBond™ Xtra — Anion exchangers featuring fast filtration, high binding capacity and high flow rate.
- Yields ultra-pure plasmid DNA in about half the time of other Midi or Maxi kits
- NucleoBond™ Xtra Midi and Maxi kits contain enlarged columns creating shorter silica resin beds
- Xtra Smart Rack hold up to 4 Maxi and 6 Midi columns for simultaneous processing
- Columns are held by the collar ring of the column
- Height of the column can be individually adjusted using plastic washers (included in the kit)
- NucleoBond™ Xtra Smart Rack is sold separately from the NucleoBond™ Xtra kit
Specifications
Specifications
| Product Type | Column holder/rack |
| For Use With (Application) | Plasmid DNA purification |
| For Use With (Equipment) | NucleBond™ Xtra Midi or Maxi kits |
| Quantity | 1 Ea. |
| Samples per Run | Up to 10 |
Product Title
Vous avez repéré une opportunité d'amélioration ?