missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoBond™ RNA Soil Mini kit for RNA from soil

Product Code. 15937282
Change view
Click to view available options
No. of Reactions:
2
10
50
Product Type:
RNA Kit
RNA Kit + DNA Set
Unit Size:
10 reactions
2 reactions
50 reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Product Type unitSize
15937282 2 RNA Kit + DNA Set 2 reactions
15858096 10 RNA Kit 10 reactions
15868096 50 RNA Kit 50 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15937282 Supplier Macherey-Nagel™ Supplier No. 740142.2

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Easy handling and superior speed in RNA purification from soil samples for qRT-PCR analysis, as well as parallel preparation of DNA. Complete kit uses anion exchange chromatography and gravity flow columns combined with NucleoSpin Bead Tubes Type A (included).

NucleoBond™ RNA Soil Mini kit enables efficient isolation of high quality RNA and DNA from any kind of soil or sediment sample. Most soils contain relatively low amounts of RNA, compared to DNA. Therefore, the kit is designed with proven NucleoBond™ technology to enable high sample input and and pure nucleic acids.

  • Anion exchange technology to optimize RNA yield and purity—suitable for qRT-PCR analysis
  • Combination of mechanical homogenization and chemical lysis allows processing of large samples
  • Parallel preparation of RNA and DNA in one hour*
  • Sample disruption is achieved with NucleoSpin™ Bead Tubes Type A, optimized Lysis Buffer, and Phenol:Chloroform:Isoamylalcohol
  • Optional Buffer OPT enables RNA and DNA purification from mineralic and organic soil samples

* DNA isolation requires the DNA Set for NucleoBond™ RNA Soil Mini, sold separately (man. part no. 740143.10 or 740143.50) or in a combo kit

Specifications

Format Gravity Flow Column
Isolation Technology Anion Exchange Chromatography, Gravity Flow Columns, NucleoSpin Bead Tubes
Elution Volume Elution volume: 50-100 μL
Sample Type Soil
Efficiency Binding capacity: 30 μg
For Use With (Equipment) Centrifuge for microcentrifuge tubes
For Use With (Application) RNA/DNA isolation from soil for qRT-PCR
Includes NucleoBond Soil Mini Columns, NucleoSpin Bead Tubes Type A, NucleoSpin RNA columns, Buffers
No. of Reactions 2
Product Type RNA Kit + DNA Set
Purity A260/A280: 1.5 - 2.0
Sample Size 0.25 to 0.5 g
Test Sensitivity RNA integrity number (RIN): > 7.0
Test Time 60 min/12 preps (RNA); 15 min/12 preps (DNA)
Yield 0.25 to 2.5 μg
Show More Show Less
compliance-icons
Product Identifier
  • NucleoBond RNA Soil Mini + DNA Set (2)
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Supplemental information
  • MIXTURE LIST-Contain: guanidium thiocyanide, ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.