missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoBond™ RNA Soil Midi kit for RNA from soil

Product Code. 15825224
Click to view available options
:
20 Preps
This item is not returnable. View return policy

Product Code. 15825224

Brand: Macherey-Nagel™ 740140.20

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Easy handling and superior speed for metagenomic soil analysis, MN Bead Tubes Type A included.

NucleoBond™ RNA Soil kit enables efficient isolation of high quality RNA and DNA from any kind of soil or sediment sample. Most soils contain relatively low amounts of RNA, compared to DNA. Therefore, the kit is designed with proven NucleoBond™ technology to enable high sample input and and pure nucleic acids.

  • Anion exchange technology to optimize RNA yield and purity—suitable for metagenomic studies
  • Combination of mechanical homogenization and chemical lysis allows processing of large samples
  • Parallel preparation of RNA and DNA in one hour*
  • Sample disruption is achieved with NucleoSpin™ Bead Tubes Type A, optimized Lysis Buffer, and Phenol:Chloroform:Isoamylalcohol
  • Optional Buffer OPT enables RNA and DNA purification from mineralic and organic soil samples

* DNA isolation requires DNA Set for NucleoBond™ RNA Soil, sold separately (man. part no. 740141.20)

Specifications

Format Gravity Flow Column
Isolation Technology Anion Exchange Chromatography, Gravity Flow Columns, NucleoSpin Bead Tubes
Elution Volume Elution volume: 100 μL
Sample Type Sediment, Soil
Efficiency Binding capacity: 600 μg
For Use With (Equipment) Centrifuge for microcentrifuge tubes
For Use With (Application) RNA isolation from soil for metagenomic study
Includes NucleoBond Soil Columns, NucleoSpin Bead Tubes Type A, NucleoSpin Finisher, buffers
No. of Reactions 20
Purity A260/A280: 1.7 - 2.1
Sample Size <2 g
Test Sensitivity RNA integrity number (RIN): > 8.5
Test Time 60 min. per 12 preps
Yield 1 - 10 μg
Show More Show Less
compliance-icons
Product Identifier
  • NucleoBond RNA Soil (20)
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contains : guanidium thiocyanide, ethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.