missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoBond™ PC EF, kit for endotoxin-free plasmid DNA

Product Code. 12860661
Change view
Click to view available options
No. of Reactions:
5
10
Yield:
100–500μg
2,000–10,000μg
500–2,000μg
Sample Volume:
30 - 150mL
150 - 500mL
500 - 2000mL
Test Time:
100 min./2 preps
150 min./2 preps
180 min./2 preps
Unit Size:
10 reactions
5 preps
5 reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Yield Sample Volume Test Time unitSize
12860661 5 500–2,000μg 150 - 500mL 150 min./2 preps 5 reactions
12863931 10 100–500μg 30 - 150mL 100 min./2 preps 10 reactions
12873971 5 2,000–10,000μg 500 - 2000mL 180 min./2 preps 5 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12860661 Supplier Macherey-Nagel™ Supplier No. 740549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Removes endotoxins via an additional washing step, without ice incubation. Macherey-Nagel™ NucleoBond™ PC EF Kit is a classic anion exchanger for purification of endotoxin-free plasmid DNA. The kit includes NucleoBond™ AX Columns, all buffers, RNase A and NucleoBond™ Filters.

An important prerequisite for transfections and gene therapy applications is endotoxin-free plasmid DNA. The patented procedure of NucleoBond™ PC EF reduces endotoxins to a very low level using the same working procedure as the standard protocol. In comparison to competitive kits, there is no need for a time-consuming incubation step to remove the endotoxins.

Features

  • Fast and easy clarification by NucleoBond™ Folded Filters—No centrifugation required, no shearing forces
  • Proven anion-exchange technology—Yields transfection-grade high-copy plasmid DNA from E. coli
  • NucleoBond™ Columns contain NucleoBond™ AX Silica Resin packed between inert filters
  • Polypropylene columns are resistant to organic solvents and are free of DNase and RNase
  • AX Resin can be used over a wide pH range and can remain in contact with buffers for up to three hours

Specifications

  • DNA with less than 0.1 EU/μg DNA
  • Purity of plasmid DNA: A260/A280 = 1.80–1.95
  • Recovery of plasmid DNA >90%
  • Structural integrity >90%
  • Yield between 500μg up to 10mg, depending on kit used

Specifications

Concentration Endotoxin level: <0.1 EU/μg
Format Mega Gravity-flow Column
Isolation Technology Anion Exchange Chromatography Technology
Efficiency Recovery of plasmid DNA: >90%
For Use With (Application) High-copy plasmids from E. coli
Includes NucleoBond AX 2000 Columns, buffers, RNase A, NucleoBond Bottle Top Filters
No. of Reactions 5
Product Type NucleoBond™ PC 2,000 EF
Quantity 1 Kit
Purity A260/A280: 1.80–1.95
RNase Yes
Sample Volume 150 - 500mL
Size <300 kbp
Test Time 150 min./2 preps
Yield 500–2,000μg
Show More Show Less
compliance-icons compliance-icons compliance-icons
Hazard Category
  • Flammable liquid Category 3
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Respiratory sensitiser Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.