missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoBond™ PC 500, Maxi Kit for Transfection-grade Plasmid DNA
Description
Features
- The classic anion exchanger for purification of transfection-grade plasmid DNA
- Fast and easy clarification of lysates with NucleoBond™ Folded Filters, no centrifugation required, no shearing forces
- NucleoBond™ Finalizers available as separate tool to avoid time consuming centrifugation for plasmid precipitation
- Proven anion-exchange technology – transfection-grade plasmid DNA
- Separate binding columns of NucleoBond™ PC kits available as NucleoBond™ AX columns
Applications:
- High-copy plasmids from E. coli
- Low-copy plasmids from E. coli (increased buffer volumes required)
Specifications
Specifications
| Format | Maxi |
| Isolation Technology | Anion Exchange Chromatography Technology |
| Final Product Type | Columns PC 500 |
| For Use With (Application) | High-copy/low-copy plasmids from E. coli |
| Includes | AX 500 Column, Buffers, RNase A, NucleoBond Folded Filters XL |
| No. of Reactions | 10 Preps |
| Product Type | Anion exchange column |
| Sample Volume | 30–150mL (HC) 100–500mL (LC) |
| Size | <300 kbp |
| Test Time | Prep time: 80–90 min./4 preps |
| Show More |
Sicherheit und Handhabung
Signalwort
- Danger
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- MIXTURE LIST-Contain: ethanol (diluted < 20 %)/sodium hydroxide solution (diluted< 2 %)/dodecyl sulfate, sodium salt
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?