missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoBond™ CB 20/CB 100, Midi kit for high integrity DNA from cells and blood
Click to view available options
Yield:
100 μg
5-20 μg
Column Type:
NucleoBond AXG 100
NucleoBond AXG 20
Unit Size:
20 reactions
Description
Genomic DNA quality is extraordinarily high. Samples are first lysed under special conditions for removal of erythrocytes. After incubation with proteinase K, the sample is diluted with buffer N2 before loading onto the NucleoBond™ column. After elution with high-salt buffer the genomic DNA is precipitated with isopropanol. The DNA purified by this method is suitable for all kinds of subsequent reactions. Kit components: NucleoBond™ CB 100 kit - NucleoBond AXG 100 columns, buffers, proteinase K. NucleoBond™ CB 20 kit - NucleoBond™ AXG 20 columns, buffers, proteinase K.
- Anion exchange technology
- Format: gravity flow columns
- Sample material: whole blood, buffy coat, cultured cells (up to 20 mL blood)
- Fragment size: 500bp to 300kb
- Yield: 100μg or 5-20μg depending on column used
- Binding capacity: 100μg or 5-20μg depending on column used
- Preparation time: 4 to 5hrs For purification of up to 500μg genomic DNA from whole blood.
Specifications
Specifications
| Column Type | NucleoBond AXG 20 |
| Format | Mini Gravity-flow Column |
| Isolation Technology | Anion Exchange Chromatography Technology |
| For Use With (Application) | Applications: Genomic DNA from whole blood, serum, plasma, cultured cells, buffy coat. |
| No. of Reactions | 20 preps |
| Product Type | NucleoBond™ CB |
| Sample Volume | 0.1-1 mL whole blood or plasma, <50 μL buffy coat, 5 x 106 cultured cells |
| Size | 500kbp |
| Test Time | 4-5hr. |
| Yield | 5-20 μg |
Safety and Handling
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction