missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Nucleobindin 1 Recombinant Rabbit Monoclonal Antibody (HL2532)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556817
This item is not returnable.
View return policy
Description
Nucleobindin 1 Recombinant Monoclonal Antibody for ICC/IF, IHC (P), Western Blot
Nucleobindin 1 encodes a member of a small calcium-binding EF-hand protein family. The encoded protein is thought to have a key role in Golgi calcium homeostasis and Ca(2+)-regulated signal transduction events.
Specifications
| Nucleobindin 1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q02818, Q02819, Q63083 | |
| NUCB1 | |
| Recombinant fragment of human Nucleobindin 1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| HL2532 | |
| Unconjugated | |
| NUCB1 | |
| B230337F23Rik; Bone 63 kDa calcium-binding protein; C77483; CALNUC; MTEST82; NUC; Nucb; Nucb1; nucleobindin 1; nucleobindin-1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 18220, 4924, 84595 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction