missing translation for 'onlineSavingsMsg'
Learn More
Learn More
nucleobindin 1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10228-1-AP-150UL
This item is not returnable.
View return policy
Description
Nucleobindin 1 encodes a member of a small calcium-binding EF-hand protein family. The encoded protein is thought to have a key role in Golgi calcium homeostasis and Ca(2+)-regulated signal transduction events.Specifications
| nucleobindin 1 | |
| Polyclonal | |
| Unconjugated | |
| NUCB1 | |
| CALNUC, NUC, NUCB1, nucleobindin 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4924 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q02818 | |
| NUCB1 | |
| nucleobindin 1 Fusion Protein Ag0290 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction