Learn More
Description
The integrated collection system incorporates a redesigned DNA collection swab and collection chamber with the established chemically-treated NUCLEIC-CARD matrix for cell lysis and protein denaturation. With this integrated collection system, the ability to achieve a high first pass success rate for reference samples and decrease laboratory costs is within reach.
Key features include:
• Higher sample transfer—redesigned N6FLOQSwabs lollipop swab and sample collection chamber for improved sample uniformity and transfer efficiency
• Certified free of DNase, RNase, and amplifiable human DNA
• Consistent, high quality STR profiles with Applied Biosystems direct PCR amplification kits
• Enables long-term archiving of samples at room temperature
• Efficient—card includes an area for cleaning strikes while punching with manual or automated puncher
• Facilitates automated processing—a color change on the card indicates the location of the sample on the matrix, helping guide commercially-available automated punching systems, as well as manual punching
• Customizable—NUCLEIC-CARD Collection Device can be purchased separately or can be included in a custom buccal sample collection kit as part of our customization services
• Easier sample collection—user-friendly instructions included in the packaging
The NUCLEIC-CARD Sample Collection Device is manufactured by Copan, a market leader in the area of sample collection tools, and distributed globally by Thermo Fisher Scientific.
NUCLEIC-CARD is a trademark of Copan Flock Technologies.
Specifications
Specifications
| Starting Material | DNA |
| Quantity | 50 Pieces |
| Product Line | NUCLEIC-CARD |
| Product Type | Collection Device System |
| Shipping Condition | Approved for shipment at Room Temperature |
| Content And Storage | 50 Pc. / case Store (at room temperature.) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.