missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Nuclease S1 (A. oryzae)
Used for Enzyme
Brand: Abnova™ P5323.10000Units
Additional Details : Weight : 0.00010kg
Description
#REF!Specifications
Enzyme Activity | |
∽34kDa | |
Native protein chromatographically purified from Aspergillus oryzae | |
Store at -20°C. | |
NUCLEASE | |
≥100,000 to 500,000 units per ml. One Unit hydrolyzes one microgram of denatured calf thymus DNA per minute at 37°C, pH 4.6. | |
None | |
Liquid |
In 30mM sodium acetate, pH 4.6, 50mM NaCl, 1mM ZnCl | |
Nuclease S1 (A. oryzae) | |
10000 Units | |
RUO | |
S1 Nuclease Assay; Eliminate non-annealed polynucleotide tails and hair-pin loops in DNA-RNA or DNA-DNA duplexes in hybridization studies and in genetic recombination experiments. | |
Fungi | |
Native protein chromatographically purified from Aspergillus oryzae |