Learn More
Nuclear Membrane Monoclonal Antibody (IPO-38), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA17628
Code : LD
Additional Details : Weight : 0.01000kg
Description
MA1-7628 detects Nuclear protein from human samples. MA1-7628 has been successfully used in ELISA procedures. The MA1-7628 immunogen is a proliferating Harvey cell line. Short term storage is at 4oC. For extended storage, aliquot and store at -80oC.
Nuclear Membrane Monoclonal antibody specifically detects Nuclear Membrane in Human samples. It is validated for ELISA, Western Blot.Specifications
Nuclear Membrane | |
Monoclonal | |
1 mg/mL | |
PBS with 0.01% sodium azide; pH 7.2 | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
Human | |
Antibody | |
IgM |
ELISA, Western Blot | |
IPO-38 | |
Unconjugated | |
Histone H1 | |
Proliferating Harvey cell line. | |
100 μg | |
Primary | |
4°C short term, -80°C long term | |
Liquid |