Learn More
Nuclear Membrane Monoclonal Antibody (1415-1 (21NC85)), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA183365
Code : LD
Additional Details : Weight : 0.01000kg
Description
Clone 1415-1 recognizes a primary band between 30 and 32kDa under non-reducing conditions. Other bands may also be present. This antibody stains nuclei of all human cell types and also stains chromosomes diffusely in metaphase cells. Intense diffuse staining is obtained in interphase cells. Human tonsil tissue can serve as a positive control for immunohistochemical applications.
Nuclear Membrane Monoclonal antibody specifically detects Nuclear Membrane in Human, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Specifications
Nuclear Membrane | |
Monoclonal | |
Unconjugated | |
Histone H1 | |
Native protein. | |
100 μL | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
1415-1 (21NC85) | |
PBS with 0.1% sodium azide | |
Mouse | |
protein A and ion exchange chromatography | |
RUO | |
Human, Rat | |
Antibody | |
IgG2a |