missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nuclear Membrane Marker Antibody (NM97), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34696F
Additional Details : Weight : 0.00970kg
Description
Nuclear Membrane Marker Monoclonal specifically detects Nuclear Membrane Marker in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
Nuclear Membrane Marker | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
Nuclei of myeloid leukemia biopsy cells | |
0.1 mL | |
Primary | |
Human | |
IgG1 κ |
Immunocytochemistry, Immunofluorescence | |
NM97 | |
Immunocytochemistry/Immunofluorescence | |
Mouse | |
Protein A or G purified | |
RUO | |
This monoclonal antibody is part of a new panel of reagents, which recognizes subcellular organelles or compartments of human cells. These markers may be useful in identification of these organelles in cells, tissues, and biochemical preparations. It recognizes an antigen associated with the nuclear membrane expressed in human cells. It can be used to stain the nuclear membrane in cell or tissue preparations and can be used as a marker of the nuclear membrane in subcellular fractions. It produces a ring pattern around the nucleus of cells of normal and malignant cells and may be used to stain the nuclear membrane of cells in fixed or frozen tissue sections. The nuclear envelope (also known as the perinuclear envelope, nuclear membrane, nucleolemma or karyotheca) is the double membrane of the nucleus that encloses genetic material in eukaryotic cells. It separates the contents of the nucleus (DNA in particular) from the cytosol (cytoplasm). Numerous nuclear pores are present on the nucl | |
Store at 4°C in the dark. |