missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nuclear Membrane Marker Antibody (NM97), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34696DL594
This item is not returnable.
View return policy
Description
Nuclear Membrane Marker Monoclonal antibody specifically detects Nuclear Membrane Marker in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence
Specifications
| Nuclear Membrane Marker | |
| Monoclonal | |
| DyLight 594 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| NM97 | |
| 50mM Sodium Borate | |
| Nuclei of myeloid leukemia biopsy cells | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction