missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Nuclear Membrane Marker Monoclonal antibody specifically detects Nuclear Membrane Marker in Human samples. It is validated for Immunofluorescence
Specifications
Specifications
| Antigen | Nuclear Membrane Marker |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Clone | AE-5 |
| Conjugate | FITC |
| Dilution | Immunocytochemistry/Immunofluorescence |
| Formulation | PBS |
| Host Species | Mouse |
| Immunogen | Nuclei of myeloid leukemia biopsy cells |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?